Skip to Content
Merck
All Photos(1)

Key Documents

25001

Sigma-Aldrich

Aluminum acetate, basic

purum, 12.5-18.4% Al basis, powder

Synonym(s):

Aluminum diacetate hydroxide, Aluminum hydroxide acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(OH)(C2H3O2)2
CAS Number:
Molecular Weight:
162.08
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

purum

form

powder

concentration

12.5-18.4% Al

impurities

≤0.002% Heavy metals (as Pb)
≤0.05% Insoluble in HCl
≤2% Ca and Mg (as (Mg)

pH

4.0-5.0

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤20000 mg/kg

cation traces

Fe: ≤200 mg/kg
Na: ≤1%

suitability

complies for appearance of solution

SMILES string

CC(=O)O[Al](O)OC(C)=O

InChI

1S/2C2H4O2.Al.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+3;/p-3

InChI key

HQQUTGFAWJNQIP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service