Skip to Content
Merck
All Photos(1)

Documents

21188

Supelco

Calcium ionophore I - membrane A

Selectophore

Synonym(s):

Calcium ionophore I - cocktail A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

product line

Selectophore

form

solid

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Ion-selective membrane for assaying calcium in whole blood, plasma and serum with ISE

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neutral carrier based ion-selective electrode for the determination of total calcium in blood serum.
P Anker et al.
Analytical chemistry, 53(13), 1970-1974 (1981-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service