Skip to Content
Merck
All Photos(1)

Key Documents

20942-U

Female Luer Connector, Tekmar® equivalent 14-0216-016

Female Luer connector, Kel-F (CTFE) fitting, thread for 1/4-28, Hamilton Tekmar® equivalent 14-0216-016

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

Kel-F (CTFE) fitting

feature

Use to connect syringes to Hamilton valves.

manufacturer/tradename

Hamilton Tekmar® equivalent 14-0216-016

packaging

1 ea of

port diam.

1.5 mm

fitting

thread for 1/4-28

Legal Information

Tekmar is a registered trademark of Tekmar Co.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service