Skip to Content
Merck
All Photos(1)

Documents

19913

Sigma-Aldrich

O-(tert-Butyldimethylsilyl)hydroxylamine

technical, ≥90% (GC)

Synonym(s):

Aminoxy-tert-butyldimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2ONH2
CAS Number:
Molecular Weight:
147.29
Beilstein:
2424306
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

bp

87-90 °C/40 mmHg (lit.)

mp

62-65 °C (lit.)
62-65 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)[Si](C)(C)ON

InChI

1S/C6H17NOSi/c1-6(2,3)9(4,5)8-7/h7H2,1-5H3

InChI key

SSUCKKNRCOFUPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Protected hydroxylamine useful for the oximation of acid- and base-sensitive compounds

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.E. Denmark et al.
The Journal of Organic Chemistry, 49, 798-798 (1984)
The Journal of Organic Chemistry, 52, 877-877 (1987)
S.E. Denmark et al.
The Journal of Organic Chemistry, 49, 4741-4741 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service