Skip to Content
Merck
All Photos(1)

Key Documents

19482

Sigma-Aldrich

Butylamine

puriss., ≥99.5% (GC)

Synonym(s):

1-Aminobutane, n-Butylamine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)3NH2
CAS Number:
Molecular Weight:
73.14
Beilstein:
605269
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.5 (vs air)

vapor pressure

68 mmHg ( 20 °C)

grade

puriss.

Assay

≥99.5% (GC)

autoignition temp.

594 °F

expl. lim.

9.8 %

impurities

≤0.2% water

refractive index

n20/D 1.401 (lit.)
n20/D 1.401

bp

78 °C (lit.)

mp

−49 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javad Gholami et al.
Luminescence : the journal of biological and chemical luminescence, 31(1), 229-235 (2015-05-29)
An N-butylamine functionalized graphene oxide nanolayer was synthesized and characterized by ultraviolet (UV)-visible spectrometry, Fourier transform infrared spectroscopy, X-ray photoelectron spectroscopy, and transmission electron microscopy. Detection of iron(III) based on photoluminescence spectroscopy was investigated. The N-butylamine functionalized graphene oxide was
D M N M Dissanayake et al.
Nanotechnology, 20(19), 195205-195205 (2009-05-08)
Organic-inorganic hybrid heterojunctions have potential as the basis for future photovoltaic devices. Herein, we report the results of investigations exploring the possibility of using pentacene and tetracene as photoelectron donors in conjunction with PbS nanocrystals (PbS-NCs). Photoinduced charge transfer was
Enhanced photovoltaic performance in nanoimprinted pentacene-PbS nanocrystal hybrid device.
Dissanayake DMNM, et al.
Applied Physics Letters, 92(9), Enhanced photovoltai-Enhanced photovoltai (2008)
Photoluminescence of CdSe Nanoparticles in the Presence of a Hole Acceptor:
n-Butylamine
Landes C, et al.
The Journal of Physical Chemistry B, 105(15), 2981-2986 (2001)
Highly efficient near-infrared hybrid organic-inorganic nanocrystal electroluminescence device.
Bourdakos KN, et al.
Applied Physics Letters, 92(15) (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service