Skip to Content
Merck
All Photos(1)

Key Documents

17037

Supelco

N-Dansyl-N′-ethylthiourea

for HPLC derivatization, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N3O2S2
CAS Number:
Molecular Weight:
337.46
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

grade

derivatization grade ((HPLC))
for HPLC derivatization

Assay

≥99.0% (HPLC)
≥99.0%

mp

160-162 °C

SMILES string

CCNC(=S)NS(=O)(=O)c1cccc2c(cccc12)N(C)C

Other Notes

Reagent for the fluorometric assay of heavy metal ios by RP HPLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Schuster et al.
Analytical and bioanalytical chemistry, 356(5), 326-330 (1996-10-01)
N-Dansyl-N'-ethylthiourea (DET) forms fluorescent chelates with a large number of heavy metal ions. The influence of the pH-value on the luminescence of DET and its metal chelates was investigated. The addition of Cu(II) to DET causes a bathochromic shift of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service