Skip to Content
Merck
All Photos(1)

Key Documents

16658

Sigma-Aldrich

X-GalNAc

reagent for selection of recombinant bacterial clones

Synonym(s):

5-Bromo-4-chloro-3-indolyl 2-acetamido-2-deoxy-β-D-galactopyranoside, 5-Bromo-4-chloro-3-indolyl N-acetyl-β-D-galactosaminide, X-Galactosaminide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18BrClN2O6
CAS Number:
Molecular Weight:
449.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for molecular biology

sterility

non-sterile

Assay

≥98.0% (TLC)

form

powder

mp

≥220 °C (lit.)

solubility

DMF: soluble
DMSO: soluble

suitability

suitable for β-galactosidase test

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]1Oc2c[nH]c3ccc(Br)c(Cl)c23

InChI

1S/C16H18BrClN2O6/c1-6(22)20-13-15(24)14(23)10(5-21)26-16(13)25-9-4-19-8-3-2-7(17)12(18)11(8)9/h2-4,10,13-16,19,21,23-24H,5H2,1H3,(H,20,22)/t10-,13-,14+,15-,16-/m1/s1

InChI key

SUWPNTKTZYIFQT-LMXXTMHSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Bromo-4-chloro-3-indolyl N-acetyl-β-D-galactosaminide (X-GalNAc)is a chromogenic substrate for for N-acetyl-β-galactosidase, used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates.

Application

Suitable for use in selection of recombinant bacterial colonies with the lac+ phenotype. Also suitable for the detection of Candida albicans.

Principle

When X-GalNAc is hydrolyzed by β-galactosidase, the resulting product will form a blue precipitate. Lac+ colonies grown in the presence of X-GalNAc turn blue in color, allowing for easy differentiation between lac+ and lac- colonies.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service