Skip to Content
Merck
All Photos(1)

Key Documents

15918

Supelco

Pyrocatechol Violet

p.a., colorimetric reagent

Synonym(s):

Catechol violet, Pyrocatecholsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14O7S
CAS Number:
Molecular Weight:
386.38
Beilstein:
353938
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

colorimetric reagent
p.a.

technique(s)

titration: suitable

mp

185 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL, clear, red
0.1 M acetate buffer pH 4.7: 10 mg/mL, clear, yellow (without a tinge of green)

SMILES string

Oc1ccc(cc1O)C(=C2\C=CC(=O)C(O)=C2)\c3ccccc3S(O)(=O)=O

InChI

1S/C19H14O7S/c20-14-7-5-11(9-16(14)22)19(12-6-8-15(21)17(23)10-12)13-3-1-2-4-18(13)27(24,25)26/h1-10,20,22-23H,(H,24,25,26)/b19-12+

InChI key

HGPSVOAVAYJEIJ-XDHOZWIPSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

λmax. 444 nm, E(1%,1 cm) ≥435 (Methylalkohol)

Other Notes

Very sensitive reagent for the determination of Fe(III); Determination of Al(III)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Abe et al.
Analytica Chimica Acta, 226, 137-137 (1989)
A. Tapparo et al.
Analytica Chimica Acta, 238, 279-279 (1990)
O. Royset
Analytica Chimica Acta, 185, 75-75 (1986)
Y. Shijo
Bulletin of the Chemical Society of Japan, 50, 1013-1013 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service