Skip to Content
Merck
All Photos(1)

Key Documents

14350

Sigma-Aldrich

Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer

purum, ≥98.0% rhodium (Rh) basis

Synonym(s):

2,5-Norbornadiene-rhodium(I) chloride dimer, Chloro(2,5-norbornadiene)rhodium(I) dimer, Rhodium(I) chloride 2,5-Norbornadiene complex dimer, [Rh(nbd)Cl]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16Cl2Rh2
CAS Number:
Molecular Weight:
460.99
EC Number:
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

grade

purum

Assay

≥98.0% rhodium (Rh) basis

storage temp.

2-8°C

SMILES string

Cl[Rh].Cl[Rh].[CH]1[CH]C2[CH][CH]C1C2.[CH]3[CH]C4[CH][CH]C3C4

InChI

1S/2C7H8.2ClH.2Rh/c2*1-2-7-4-3-6(1)5-7;;;;/h2*1-4,6-7H,5H2;2*1H;;/q;;;;2*+1/p-2/t2*6-,7+;;;;

InChI key

RXDWVIOULPVOEO-SEGRDSFWSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

Rhodium(I) complex used for the preparation of Rh-phosphine complexes [1]; Catalyst used for the [2+2]-cycloreversion of strained cyclobutenes [2][3]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Hayashi et al.
Journal of the American Chemical Society, 109, 7876-7876 (1987)
W. Marterer et al.
Tetrahedron Letters, 28, 5497-5497 (1987)
L.A Paquette et al.
Journal of the American Chemical Society, 108, 7431-7431 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service