09243
4-Aminophenyl-β-D-thiogalactopyranoside-Agarose 4B
matrix 4% beaded agarose
Synonym(s):
APTG-Agarose
About This Item
Recommended Products
General description
Application
Physical form
Other Notes
Disclaimer
![](/deepweb/assets/sigmaaldrich/product/images/242/317/89e134be-e488-4e6d-9500-3baa2929e5c3/300/89e134be-e488-4e6d-9500-3baa2929e5c3.jpg)
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service