Skip to Content
Merck
All Photos(1)

Documents

08720

Sigma-Aldrich

4-Amino-5-hydroxy-2,7-naphthalenedisulfonic acid disodium salt

technical, 65-80% (T)

Synonym(s):

8-Amino-1-naphthol-3,6-disulfonic acid disodium salt, Disodium 4-amino-5-hydroxy-2,7-naphthalenedisulfonate, H Acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H4(OH)(SO3Na)2
CAS Number:
Molecular Weight:
363.27
Beilstein:
3863707
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

technical

Assay

65-80% (T)

impurities

monosodium salt
10-20% Na2SO4

SMILES string

[Na+].[Na+].Nc1cc(cc2cc(cc(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service