Skip to Content
Merck
All Photos(1)

Key Documents

07442

Supelco

D-(+)-Turanose

analytical standard

Synonym(s):

3-O-α-D-Glucopyranosyl-D-fructose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93771
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥98% (GC)

optical activity

[α]20/D 75.8±1.0°, c = 4 in H2O

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

oligosaccharides

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](O[C@@H]2[C@@H](O)[C@@H](O)COC2(O)CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-5-7(17)8(18)9(19)11(22-5)23-10-6(16)4(15)2-21-12(10,20)3-14/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12?/m1/s1

InChI key

SEWFWJUQVJHATO-PUVWEJBASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Turanose is an analog of sucrose that is not metabolized by higher plants but is used as a carbon source by many bacteria, fungi, and other organisms. It is taken up into plant cells by sucrose transporters and is involved in intracellular sugar signaling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Torsten Juelich et al.
Journal of immunology (Baltimore, Md. : 1950), 183(11), 7063-7072 (2009-11-17)
The role of chromatin remodeling and histone posttranslational modifications and how they are integrated to control gene expression during the acquisition of cell-specific functions is poorly understood. We show here that following in vitro activation of CD4(+) and CD8(+) T
Noa Dahan-Pasternak et al.
Journal of cell science, 126(Pt 14), 3055-3069 (2013-05-21)
In Plasmodium falciparum, the deadliest form of human malaria, the nuclear periphery has drawn much attention due to its role as a sub-nuclear compartment involved in virulence gene expression. Recent data have implicated components of the nuclear envelope in regulating
Felix Lohmann et al.
Stem cells (Dayton, Ohio), 28(2), 201-212 (2009-12-17)
Dynamic regulation of histone methylation by methyltransferases and demethylases plays a central role in regulating the fate of embryonic stem (ES) cells. The histone H3K9 methyltransferase KMT1E, formerly known as ESET or Setdb1, is essential to embryonic development as the
Gabi Gerlitz et al.
Traffic (Copenhagen, Denmark), 8(11), 1521-1529 (2007-09-08)
Directed cell migration is a property central to multiple basic biological processes. Here, we show that directed cell migration is associated with global changes in the chromatin fiber. Polarized posttranslational changes in histone H1 along with a transient decrease in
Nikki R Kong et al.
Cell reports, 34(1), 108574-108574 (2021-01-07)
The zinc finger transcription factor SALL4 is highly expressed in embryonic stem cells, downregulated in most adult tissues, but reactivated in many aggressive cancers. This unique expression pattern makes SALL4 an attractive therapeutic target. However, whether SALL4 binds DNA directly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service