Skip to Content
Merck
All Photos(1)

Key Documents

00373

Supelco

4-Methylbenzophenone

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COC6H5
CAS Number:
Molecular Weight:
196.24
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

326 °C (lit.)

mp

56.5-57 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical

format

neat

SMILES string

Cc1ccc(cc1)C(=O)c2ccccc2

InChI

1S/C14H12O/c1-11-7-9-13(10-8-11)14(15)12-5-3-2-4-6-12/h2-10H,1H3

InChI key

WXPWZZHELZEVPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methylbenzophenone is a photo-initiator for printing inks, mainly applied to the cardboard boxes used for packing. It is reported to be a non-genotoxic carcinogen. However, it can contaminate foodstuffs when it comes in contact with the package contents.

Application

4-Methylbenzophenone may be used as a reference standard for the determination of the analyte in breakfast cereals, paperboard food packaging and packed milk by various chromatography techniques.

Analysis Note

The purity is calculated based on the principle of mass balance and is mentioned on the CoA

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A safety evaluation of printed paper and board contaminants: Photo-initiators as a case study
Van HE, et al.
Reference Module in Life Sciences, 1-13 (2017)
Solid-phase microextraction followed by gas chromatography-mass spectrometry for the determination of ink photo-initiators in packed milk
Negreira N, et al.
Talanta, 82(1), 296-303 (2010)
Analysis of benzophenone and 4-methylbenzophenone in breakfast cereals using ultrasonic extraction in combination with gas chromatography-tandem mass spectrometry (GC-MSn)
Van HE, et al.
Analytica Chimica Acta, 663(1), 55-59 (2010)
Rapid multi-analyte quantification of benzophenone, 4-methylbenzophenone and related derivatives from paperboard food packaging
Koivikko R, et al.
Food Additives and Contaminants, 27(10), 1478-1486 (2010)
4-Methylbenzophenone and benzophenone are inactive in the micronucleus assay
Abramsson-Zetterberg L and Svensson K
Toxicology Letters, 201(3), 235-239 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service