Skip to Content
Merck
All Photos(1)

Documents

21-1370

Sigma-Aldrich

Nickel(II) sulfate hexahydrate

SAJ first grade, 98.5-102.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 6H2O
CAS Number:
Molecular Weight:
262.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

vapor density

2.07 (vs air)

Assay

98.5-102.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: nickel

availability

available only in Japan

SMILES string

O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2

InChI key

RRIWRJBSCGCBID-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Mattarozzi et al.
Micron (Oxford, England : 1993), 75, 18-26 (2015-05-20)
Environmental scanning electron microscopy (ESEM) permits to analyze samples in their native-hydrated state, allowing a broad spectrum of biological applications. In this study, ESEM equipped with energy dispersive X-ray spectrometer (EDS) was used as a fast method to analyze tissue
Xiaohua Xu et al.
Particle and fibre toxicology, 9, 40-40 (2012-11-07)
It has been well recognized that toxicity of fine ambient air particulate matter (PM(2.5)) may depend on its chemical constituents, including components such as soluble metals that may theoretically exert distinctive effects. We have recently demonstrated an important effect of
A Tammaro et al.
International journal of immunopathology and pharmacology, 25(2), 519-521 (2012-06-16)
Nickel deficiency leads to reduced iron content in organs and to reduced hemoglobin levels and haematocrit counts. We carried out a clinical trial of oral hyposensitization therapy with low doses of nickel on a group of 20 patients affected by
K Amudha et al.
Chemico-biological interactions, 193(1), 57-64 (2011-05-24)
This study was conducted to investigate the beneficial role of naringin on nickel induced nephrotoxicity. Nickel (Ni) (20mg/kg body weight (b.w.) was administered intraperitoneally (i.p.) for 20 days. Naringin was administered orally (20, 40 and 80 mg/kg b.w.) with i.p.
Charlotte G Mortz et al.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology, 24(1), 84-87 (2013-01-22)
We have previously reported patch test reactivity to nickel sulphate in a cohort of unselected infants tested repeatedly at 3-18 months of age. A reproducible positive reaction at 12 and 18 months was selected as a sign of nickel sensitivity provided a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service