Skip to Content
Merck
All Photos(1)

Key Documents

11140957001

Roche

GTP

=98% (HPLC), solution, pkg of 400 μL (100 mM; 40 μmol)

Synonym(s):

GTP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

description

Lithium salt

Assay

98% (HPLC)

form

solution

packaging

pkg of 400 μL (100 mM; 40 μmol)

manufacturer/tradename

Roche

storage temp.

−20°C

General description

GTP is a 100 mM solution of the lithium salt (pH 7).
Guanosine triphosphate (GTP) is a purine nucleoside triphosphate. This nucleotide is mostly used as substrate in RNA synthesis during transcription. It is comprised of a ribose sugar and three phosphates with nucleobase and triphosphate moiety attached to 1′and 5′ carbons. It may also be involved in signal transduction, energy transfer within the cell, as an activator of substrates in metabolic reaction or for protein synthesis and gluconeogenesis.

Application

GTP has been used in cell-free generation of COPII-coated procollagen I carriers[1] and to label transcription elongation complexes.[2]
GTP, lithium salt, is suitable for applications such as in vitro RNA transcription.

Quality

Typical analysis:
  • ≥98% GTP (HPLC), ≤1.5% GDP (HPLC), ≤0.5% GMP (HPLC).
  • Tested for the absence of RNases according to the current Quality Control procedures.

Unit Definition

1 mM GTP = 13.7 A252 units

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mfd Dynamically Regulates Transcription via a Release and Catch-Up Mechanism
Le TT, et al.
Cell (2017)
Cell-free Generation of COPII-coated Procollagen I Carriers
Yuan L, et al.
Bio-protocol, 7(22) (2017)
Youngseo Cheon et al.
Genome biology, 22(1), 294-294 (2021-10-20)
Promoter-proximal pausing of RNA polymerase II (RNAPII) is a critical step for the precise regulation of gene expression. Despite the apparent close relationship between promoter-proximal pausing and nucleosome, the role of chromatin remodeler governing this step has mainly remained elusive.
Riku Nagai et al.
Methods and protocols, 4(3) (2021-07-22)
The rates of translation elongation or termination in eukaryotes are modulated through cooperative molecular interactions involving mRNA, the ribosome, aminoacyl- and nascent polypeptidyl-tRNAs, and translation factors. To investigate the molecular mechanisms underlying these processes, we developed an in vitro translation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service