Skip to Content
Merck
All Photos(1)

Documents

ZFMQ000PR

Milli-Q

Feed Water Pressure Regulator

Regulates the feed water pressure above 0.3 bars, Adjusts inlet water pressure upstream of water purification systems to regulate the pressure according to the system′s requirements

Synonym(s):

Feed Water Pressure Regulator for Milli-Q® Systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JA.14

material

stainless steel valve body (with EPDM molding)

packaging

pkg of 1 unit

L × W × H

165 mm × 130 mm × 125 mm

compatibility

for use with Synergy® UV
for use with Synergy®

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Adjusts inlet water pressure upstream of water purification systems to regulate the pressure according to the system′s requirements.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SYNERGY is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service