WBAVDCH01
Immobilon® Block - CH (Chemiluminescent Blocker)
Chemiluminescent detection kit used for Western blotting, dot/slot blotting
Synonym(s):
Western blot blocking buffer, blocking buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
conjugate
peroxidase conjugate
Quality Level
packaging
pkg of 500 mL
manufacturer/tradename
Immobilon®
technique(s)
western blot: suitable
membrane area
1000 cm2 , membrane coverage
detection method
chemiluminescent
shipped in
ambient
storage temp.
15-25°C
Related Categories
General description
Immobilon® Block Noise Cancelling Reagents are designed especially for Western blotting applications. This family of protein-free, ready-to-use buffers has been optimized to reduce background levels when using chemiluminescence, fluorescent, or phosphotyrosine detection. Bløk buffers are ideal for use with Immobilon-P Western blotting membranes and the SNAP i.d. system, and are also compatible with standard Western blotting techniques.
Application
For use on immunoblots.
Packaging
Sufficient reagent for 19 mini blots (8x7 cm)
Quality
For optimum results, use Immobilon-P membrane for chemiluminescence (luminol) or visual detection (e.g. TMB), and Immobilon-FL membrane for fluorescent detection applications.
Storage and Stability
Stable for 2 years at 20–25°C
Legal Information
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biochimie, 94(12), 2639-2648 (2012-08-21)
Isoprenoids are recognized for their ability to suppress carcinogenic processes in vivo and in vitro. We previously established that the isoprenoid, perillyl alcohol, acted mechanistically on translation of specific proteins through modulation of mechanistic target of rapamycin (mTOR) signaling. Telomerase-the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service