Skip to Content
Merck
All Photos(1)

Key Documents

SCVPU02RE

Millipore

Stericup-VP Sterile Vacuum Filtration System

250 mL process volume, 0.1 µm pore size, polyethersulfone membrane, pkg of 12, radio-sterilized

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.22

material

polyethersulfone membrane
polystyrene

sterility

sterile

feature

holdup volume 2.4 mL
hydrophilic

manufacturer/tradename

Stericup®

parameter

250 mL process volume
250 mL sample volume
45 °C max. temp.

technique(s)

sterile filtration: suitable

H

164 mm

diam.

12.5 cm

filter diam.

73 mm

filtration area

40 cm2

funnel capacity

250 mL

matrix

Express PLUS

pore size

0.1 μm nominal pore size
0.1 μm pore size

fitting

inlet funnel
outlet flask (Receiver)

shipped in

ambient

General description

250 mL bottle with cap
The Stericup® vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop filter unit with a receiver flask for processing and storing volumes from 150 mL to 1,000 mL in a single, ready-to-use format. Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device. The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop bottle-top devices with your own sterile capture flask or bottle.

Features & Benefits

Millipore Express PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications

Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component

Application

250 mL process volume, 0.1 µm pore size, polyethersulfone membrane, pkg of 12, radio-sterilized

Principle

Vacuum

Physical form

Color Code: Clear with blue adapter

Preparation Note

Sterilization Method
Gamma irradiation

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Prior to probing a membrane using precious antibodies, it is helpful to visualize the transferred proteins on the blot to ensure complete transfer and even loading. This page describes possible causes and potential remedies for challenges encountered during protein visualization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service