SCT269
BioTracker™ Red Exosome Protein Labeling Kit
Human
Synonym(s):
exosome protein dye, exosome protein label, exosome protein probe, fluorescent exosome protein label, red exosome protein dye, red exosome protein label, red exosome protein probe
About This Item
Recommended Products
product name
BioTracker™ Red Exosome Protein Labeling Kit,
biological source
human
Quality Level
packaging
vial of 5 tests (reactions)
manufacturer/tradename
Millipore
technique(s)
cell culture | stem cell: suitable
flow cytometry: suitable
shipped in
2-8°C
storage temp.
2-8°C
General description
Features and Benefits
Target description
Storage and Stability
Other Notes
Legal Information
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service