SCT081
BioTracker Cysteine-Glo Live Cell Probe Antibody, BioLuminescent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥97% (HPLC)
mol wt
calculated mol wt 348.4
packaging
pkg of 50 mL
manufacturer/tradename
Millipore
storage condition
desiccated
protect from light
technique(s)
cell based assay: suitable
detection method
bioluminescent
shipped in
ambient
storage temp.
−20°C
General description
BioTracker Cysteine-Glo Live Cell Probe is a turn-on bioluminescence probe for detecting exogenous and endogenous cysteine in vitro and in vivo. Cysteine (Cys) is a semi-essential amino acid that plays an important role in numerous biological functions. The Cysteine-Glo probe is a Cys-selective, reaction-based bioluminescent probe that can sensitively monitor Cys in biological systems. This probe may be used for real-time visualization of fluctuations in Cys levels in live cells and whole animals.
Reference:
1. Hu, S., Lu, P., Zhou, S., Kang, T., Hai, A., Ma, Y., ... & Li, M. (2020). Bioluminescence imaging of exogenous & endogenous cysteine in vivo with a highly selective probe. Bioorganic & medicinal chemistry letters, 30(6), 126968.
Reference:
1. Hu, S., Lu, P., Zhou, S., Kang, T., Hai, A., Ma, Y., ... & Li, M. (2020). Bioluminescence imaging of exogenous & endogenous cysteine in vivo with a highly selective probe. Bioorganic & medicinal chemistry letters, 30(6), 126968.
Application
Purity ≥ 97% confirmed by HPLC. Identification confirmed by HNMR, LC-MS, and elemental analysis.
Features and Benefits
Turn-on bioluminescence probe that can sensitively monitor Cysteine in live cells and animal studies.
Physical form
Lyophilized. White solid.
Storage and Stability
Store at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bioorganic & medicinal chemistry letters, 30(6), 126968-126968 (2020-02-06)
Cysteine (Cys) is a semi-essential amino acid that exerts a vital role in numerous biological functions. A noninvasive method for in vivo imaging of cysteine could represent a valuable tool for research cysteine and its complex contributions in living organisms.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service