Skip to Content
Merck
All Photos(1)

Key Documents

SCC414

Sigma-Aldrich

S462 Human Peripheral Nerve Sheath Tumor Cell Line

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.28
Product SCC414 is not currently sold in your country. Contact Technical Service

Product Name

S462 Human Peripheral Nerve Sheath Tumor Cell Line,

biological source

human

packaging

pkg of >1X10⁶ cells

manufacturer/tradename

Sigma-Aldrich

growth mode

N/A

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

≤ − 140°C

General description

Malignant peripheral nerve sheath tumors (MPNSTs) are soft tissue sarcomas that arise from peripheral nerves and result in high rates of local recurrence and hematogenous metastasis. Half of MPNSTs occur in patients with neurofibromatosis type 1 (NF1), a common autosomal dominant multisystem disorder caused by mutations of the NF1 tumor suppressor gene located on chromosome 17q. Loss of function of the intact NF1 allele, due to loss of heterozygosity (LOH) and somatic mutations of the gene, is associated with dermal and plexiform neurofibroma formation. Approximately 10% of NF1 patients develop MPNST, of whom only 21% survive for five years after diagnosis.

Application

Each vial contains ≥ 1X10⁶ viable cells.Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.Cells are verified to be of human origin and negative for inter-species contamination from mouse, rat, Chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services Cells are negative for mycoplasma contamination.Each lot of cells are genotyped by STR analysis to verify the unique identity of the cell line.
Malignant peripheral nerve sheath tumors (MPNSTs) are soft tissue sarcomas that arise from peripheral nerves and result in high rates of local recurrence and hematogenous metastasis. Half of MPNSTs occur in patients with neurofibromatosis type 1 (NF1), a common autosomal dominant multisystem disorder caused by mutations of the NF1 tumor suppressor gene located on chromosome 17q. Loss of function of the intact NF1 allele, due to loss of heterozygosity (LOH) and somatic mutations of the gene, is associated with dermal and plexiform neurofibroma formation. Approximately 10% of NF1 patients develop MPNST, of whom only 21% survive for five years after diagnosis. Source S462 is an established human MPNST cell line from a clinically and genetically well-characterized NF1 patient, with a nonsense germline mutation in exon 37 of the NF1 gene, LOH of the NF1 gene, LOH of the p53 gene, and a homozygous missense mutation in exon 4 of the p53gene.As a verified MPNST cell line, S462 is a valuable tool for the further investigation of the biology and pathogenesis of this malignancy as well as for in vitro pharmacologic studies essential for the development of new therapies.

Features and Benefits

S462 Human Peripheral Nerve Sheath Tumor Cell Line is a varified MPNST cell line and a valuable tool for the further investigation of the biology and pathogenesis of this malignancy.

Storage and Stability

Cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service