Skip to Content
Merck
All Photos(1)

Key Documents

S7200-M

Sigma-Aldrich

ApopTag Peroxidase In Situ Oligo Ligation (ISOL) Kit

Sign Into View Organizational & Contract Pricing

Select a Size

40 ASSAYS
£715.00

£715.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
40 ASSAYS
£715.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

£715.00


Please contact Customer Service for Availability

Request a Bulk Order

technique(s)

immunohistochemistry: suitable

Quality Level

General description

The ApopTag Peroxidase In Situ Oligo Ligation Kit specifically detects apoptosis by staining only cells that contain double-stranded breaks that are blunt-ended or have a one base 3′ overhang (cells containing nicked, gapped, 3′-recessed, 3′-overhanging ends longer than one base and single-stranded ends are not detected). The high analytic specificity of the ApopTag ISOL Kit minimizes background due to non-specific labeling of DNA. The specificity can be exploited to more readily identify apoptosis in systems presenting cells with both apoptosis and DNA damage or apoptosis and necrosis. Results are visualized using brightfield microscopy.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service