Skip to Content
Merck
All Photos(4)

Key Documents

PD1504700

Millipore

PetriSlides

For microbial contamination analysis, Membrane filtration, pkg of (100 units)

Synonym(s):

Petri dish for membrane filtration

Sign Into View Organizational & Contract Pricing

Select a Size

100 UNITS
£120.00

£120.00


Estimated to ship on19 May 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
100 UNITS
£120.00

About This Item

UNSPSC Code:
41122101
eCl@ss:
32030209
NACRES:
NB.14

£120.00


Estimated to ship on19 May 2025Details


Request a Bulk OrderRequest more information

material

cellulose membrane
clear device
polystyrene slide

Quality Level

sterility

non-sterile

manufacturer/tradename

Petrislides

packaging

pkg of (100 units)

technique(s)

sterile filtration: suitable

input

sample(s) (environmental)

application(s)

bioburden testing
(Beverages, Water)

compatibility

Membrane filtration

shipped in

ambient

Related Categories

General description

PetriSlides are clear devices suitable for the examination of microbial contamination. They are an alternative to standard size Petri dishes and are compatible with membrane filters. The PetriSlides hold the filters securely in place. Their transparent cover allows for microscopic examination. The rectangular base with rounded corners makes it easy to mount on the microscope stage.

Application

PetriSlides are used to analyze microbiological contaminations in the water and beverage samples. They are storage and examination containers for test filters.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The Millipore® Patch Test Kit is a portable kit for the collection and testing of hydraulic fluids, bulk chemicals, boiler water, and lubricating oils. The kit includes consumables for 100 tests.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service