Skip to Content
Merck
All Photos(1)

Documents

PBQK04310

Millipore

PES Membrane Filter, 50 kDa NMWCO

Biomax®, filter diam. 44.5 mm

Synonym(s):

Biomax PES ultrafiltration membrane discs, Ultrafiltration Discs, 50 kDa NMW

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602
NACRES:
NB.24

material

polyethersulfone membrane

Quality Level

manufacturer/tradename

Biomax®

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

filter diam.

44.5 mm

filtration area

13.4 cm2

pore size

50 kDa NMWCO

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

  • Ultrafiltration cut disc membranes in Biomax® polyethersulfone (PES) are ideal for protein concentration, purification, and desalting.
  • Optimized for concentrating or desalting serum, plasma, or conditioned tissue culture media.
  • A wide range of pore sizes is available to suit your application needs.

Application

50 kDa NMWL, Biomax polyethersulfone, 44.5 mm diameter

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service