Skip to Content
Merck
All Photos(2)

Key Documents

NXBH001BEN

Millipore

Natrix® Bench Holder

new

Synonym(s):

Membrane chromatography holder

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
£118.00

£118.00


In StockDetails



Select a Size

Change View
1 UNIT
£118.00

About This Item

UNSPSC Code:
12352200

£118.00


In StockDetails


description

Holder for Natrix® Bench devices

Quality Level

product line

Natrix®

packaging

pack of 1

application(s)

pharmaceutical

storage temp.

2-30°C

General description

Holder for Natrix® Bench devices

Application

Chromatography

Biochem/physiol Actions

Equipment for purification of biomolecules

Packaging

Carton, 1/pack

Legal Information

NATRIX is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service