NE-2ML-B
Anti-M (clone LM110/140), Blood Grouping Reagent
BIOSCOT™ Anti-M from the clone LM110/140, Murine IgG, Blood Grouping Reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
clone
LM110/140, monoclonal
packaging
pkg of 2 mL
manufacturer/tradename
BIOSCOT™
storage condition
do not freeze
concentration
15-25 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
8.1-8.7
isotype
IgG
application(s)
clinical testing
shipped in
ambient
storage temp.
2-8°C
Related Categories
General description
Contains 0.1% (w/v) Sodium Azide
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months from date of filtration
CE Mark
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months from date of filtration
CE Mark
Application
For use by Tube technique
Vialled, Labeled
Legal Information
BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service