Skip to Content
Merck
All Photos(1)

Key Documents

MXRPWRSPAU

Millipore

Milliflex® Rapid System 2.0 power supply

Milliflex Oasis pumppower supply for Austria, for use with the Milliflex Rapid 2.0 Detection Tower or the Milliflex Rapid 2.0 Autospray Station

Synonym(s):

Milliflex® Rapid System 2.0 power supply

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
£212.00

£212.00


Available to ship on11 April 2025Details



Select a Size

Change View
1 EACH
£212.00

About This Item

UNSPSC Code:
41122409
NACRES:
JA.13

£212.00


Available to ship on11 April 2025Details


packaging

pkg of 1 ea

manufacturer/tradename

Milliflex®

technique(s)

Rapid Sterility Testing: suitable

application(s)

bioburden testing
pharmaceutical
sterility testing

compatibility

for use with the Milliflex Rapid 2.0 Detection Tower or the Milliflex Rapid 2.0 Autospray Station

Looking for similar products? Visit Product Comparison Guide

General description

The Milliflex® Rapid System 2.0 is an automated solution for the rapid detection, response and resolution of microbial contamination in filterable samples throughout the manufacturing process. Milliflex® Rapid System 2.0 power supply is set with of European standard with 100 - 240 V, 50/60 enables efficient functioning of the Milliflex Rapid 2.0 Detection Tower or the Milliflex Rapid 2.0 Autospray Station.

Application

Milliflex® Rapid System 2.0 finds its application in sterility testing and bioburden testing for Pharmaceutical Products Analysis.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service