Skip to Content
Merck
All Photos(3)

Documents

MIGSWG250

Millipore

Microfil® funnel with S-Pak®

sample volume 250 mL, mixed cellulose esters (MCE), pore size 0.22 μm, filter diam. 47 mm, white filter, gridded filter, for water, beverage, and pharmaceutical application.

Synonym(s):

Membrane filtration system, MCE Membrane, GSWG Membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

gridded filter
mixed cellulose esters (MCE)
polypropylene
white filter

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic

manufacturer/tradename

Microfil® with S-Pak®

parameter

250 mL sample volume

technique(s)

sterile filtration: suitable

filter diam.

47 mm

matrix

MF-Millipore

pore size

0.22 μm pore size

input

sample(s) (environmental)
sample type food(s)
sample type beverage(s)

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The Microfil® filtration system is a simple and reliable solution for routine testing of raw materials, beverages, and water for microbiological contamination. The Microfil® filtration system eliminates time-consuming steps, equipment setup, waste, and autoclaving and improves the productiveness of the lab. This cost-effective system uses pre-sterilized, ready-to-use funnels and membranes for filtration support. Ultra-thin 100 or 250 mL Microfil® funnels eliminate the need for washing and sterilizing after each test, making equipment handling easy. A range of pre-sterilized 47 mm diameter gridded membranes is available with the Microfil® funnels. Individually sealed membranes are placed in peel-back envelopes (S-Pak® membranes). Due to the membrane support, filtration rates with the Microfil® filtration system are faster when compared to traditional filtration equipment.

Application

The Microfil® filtration system enables fast and accurate microbiological analysis of various liquid samples. The filtration system is suitable for the food and beverage, pharmaceutical, and cosmetic industries.

Features and Benefits

  • Use of 47 mm gridded pre-sterilized membranes
  • Hydrophobic funnel surfaces for improved recovery
  • Smooth interior funnel surfaces with no scratches
  • Lip seal to prevent membrane by-pass
  • Food-contact-polymer funnels, free from agents that may inhibit bacterial growth
  • Pre-sterilized single use funnel can be directly used eliminating the use of Flaming
  • Recyclable plastic funnel material that allows for greater environmental protection

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service