Key Documents
MAVFA4VP2
Millipore
MAS-100 VF® Portable Air Sampler Validation Protocol
for use with MAS-100 VF® Microbial Air Samplers
Select a Size
1 EACH
£1,330.00
£1,330.00
Available to ship on10 April 2025Details
Request more information
Select a Size
Change View
1 EACH
£1,330.00
About This Item
UNSPSC Code:
41116500
NACRES:
JA.11
£1,330.00
Available to ship on10 April 2025Details
Request more information
availability
not available in North America
compatibility
for use with MAS-100 VF® Microbial Air Samplers
General description
MAS-100 VF® Validation Protocol(A4 Format) is designed as a ready to use step by step guide to perform IQ and OQ activities of MAS-100 VF® microbial air sampling systems. The protocol contains an exhaustive list of tests to cover a batch of units to be validated, the test records protocol MAVFA4TR1 including test descriptions and test records must be ordered separately. The protocol follows GMP, EP, USP and JP regulatory guidelines. Not available in North America.
Application
MAS-100 VF® Validation Protocol is used to validate microbial air sampling systems with ease and consistent performance while following regulatory guidelines.
Legal Information
MAS-100 VF is a registered trademark of MBV AG