Skip to Content
Merck
All Photos(1)

Key Documents

MABX4013

Sigma-Aldrich

Anti- Insulin-like growth factor binding protein 4 (IGFBP-4) clone 13

clone 13, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
£9,090.00

£9,090.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
£9,090.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

£9,090.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

Gene Information

human ... IGFBP4(3487)

General description

Antibody recognizes the N-term of Human insulin like growth factor binding protein 4 (IGFBP-4). IGFBP-4 is a member of the Insulin like growth factor family and is responsible for binding insulin like growth factors I and II. Research has linked levels of C-term (CT-IGFBP-4) and N-term (NT-IGFBP-4) to certain cardiac events, offering potential as a novel cardiac biomarker

Specificity

Human

Application

Confirmed matched pairing. Capture Antibody MABX4013-1KC; Detector Antibody MABX4144-1KC or Detector Antibody MABX4180-1KC
Research Category
Cell Structure
This mouse monoclonal Anti- Insulin-like growth factor binding protein 4 (IGFBP-4) clone 13, Cat. No. MABX4013-1KC is validated for use in and Western Blotting and ELISA, for the detection of NT-IGFBP-4.

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal in buffer containing PBS (pH 7.4) with 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service