Prolactin (PRL) also known as luteotropic hormone is a neuroendocrine hormone secreted by the pituitary gland. It is commonly measured as part of a sex hormone screen to investigate fertility problems. Elevated levelso of PRL induce lactation and can inhibit secretion of gonadotropins such as Follicle Stimulating Hormone (FSH) and Gonadotropin-releasing hormone (GnRH)
ELISA and Lateral Flow Immunoassay.
Physical form
Format: Purified
Other Notes
Concentration: Please refer to lot specific datasheet.
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.