Skip to Content
Merck
All Photos(1)

Key Documents

MABT1574

Sigma-Aldrich

Anti-Profilin IIa Antibody, clone PF2A1-8F8

Synonym(s):

Profilin II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

PF2A1-8F8, monoclonal

mol wt

calculated mol wt 15.05 kDa
observed mol wt ~15 kDa

purified by

using protein G

species reactivity

human, rat

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Profilin-2 (UniProt: P35080; also known as Profilin II) is encoded by the PFN2 gene (Gene ID: 5217) in human. Profilins are actin-binding proteins that serve as key regulators of actin polymerization in cells. Five profilin isoforms have been identified in mammals: Profilins I, IIa, IIb, III and IV. Profilin binds to G-actin in a 1:1 complex and thereby sequesters monomeric actin. Profilin I is highly expressed in all tissues except skeletal muscle, heart, and brain, while profilin II is highly expressed in brain, skeletal muscle, and kidney. Profilin IIa is highly expressed in developing and adult neurons and is involved in correct neurite outgrowth. Profilin IIb appears to be a minor form and it differs from IIa in 13 amino acids in the C-terminal region. Knockdown of both profilins I and II is reported to inhibit neurite outgrowth, possibly impairing actin dynamics by reducing the availability of profilin -G-actin complexes for microfilament growth. In addition to its role in actin polymerization and neurite outgrowth, profilin IIa may also be involved in regulation of synaptic vesicles. It exhibits high affinity for proline-rich peptides and is shown to interact with survival of motor neurons protein (SMN). Mutated forms of SMN (Y272G and G279V) are reported to significantly reduce its interaction with profilin IIa. (Ref.: Sharma, A., et al. (2005). Exp. Cell Res. 305; 195-197; Witke, W., et al. (1998). EMBO J. 17; 967-976).

Specificity

Clone PF2A1-8F8 is a mouse monoclonal antibody that specifically detects Profilin-II. It targets an epitope within 10 amino acids from the C-terminal half.

Immunogen

T7-tagged full-length recombinant human Profilin IIa.

Application

Quality Control Testing

Evaluated by Western Blotting in Human cerebral cortex tissue lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected Profilin IIa in Human cerebral cortex tissue lysate.

Tested Applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated Profilin IIa in Immunoprecipitation applications (Sharma, A., et al. (2005). Exp Cell Res. 309(1): 185-97).

Immunofluorescence Analysis: A representative lot detected Profilin IIa in Immunofluorescence applications (Sharma, A., et al. (2005). Exp Cell Res. 309(1): 185-97).

Western Blotting Analysis: A representative lot detected Profilin IIa in Western Blotting applications (Sharma, A., et al. (2005). Exp Cell Res. 309(1): 185-97).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Profilin IIa, clone PF2A1-8F8, Cat. No. MABT1574, is a mouse monoclonal antibody that detects Profilin IIa and is tested for use in Immunofluorescence, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service