MABS2280
Anti-CLEC18A Antibody, clone 3F10F
Synonym(s):
C-type lectin domain family 18 member A;Mannose receptor-like protein 2
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
3F10F, monoclonal
mol wt
calculated mol wt 49.6 kDa
observed mol wt ~58 kDa
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 100 μg
technique(s)
ELISA: suitable
flow cytometry: suitable
western blot: suitable
isotype
IgG1κ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
shipped in
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CLEC18A(348174)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting with recombinant Human CLEC18A protein.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant Human CLEC18A protein.
Tested Applications
ELISA Analysis: A representative lot detected CLEC18A in ELISA applications (Liao, T-L., et al. (2018). Sci Rep. 8(1); 17287).
Flow Cytometry Analysis: A representative lot detected CLEC18A in Flow Cytometry applications (Liao, T-L., et al. (2018). Sci Rep. 8(1); 17287).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service