Skip to Content
Merck
All Photos(2)

Key Documents

MABS21

Sigma-Aldrich

Anti-PORCN Antibody, clone 15G12.1

clone 15G12.1, from mouse

Synonym(s):

Probable protein-cysteine N-palmitoyltransferase porcupine, Protein MG61

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
£307.00

£307.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
£307.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

£307.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15G12.1, monoclonal

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Porcn(53627)

General description

PORCN is a multi-pass endoplasmic reticulum protein involved in the processing of wingless and other Wnt proteins that facilitate their secretion from Wnt-producing cells. Studies have shown mutations in the genes that encode for PORCN cause a pleiotropic X-linked dominant disorder, focal dermal hypolasia (FDH, also known as Goltz syndrome). Skeletal abnormalities such as syndactyly, ectordactyly, and brachydactyly, and some cases of osteopathia striata have also been attibuted to PORCN abnormalities.

Immunogen

Linear peptide corresponding to human PORCN.

Application

Anti-PORCN Antibody, clone 15G12.1 is an antibody against PORCN for use in WB & IC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected PORCN in HeLa and A431 cells.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected PORCN in 10 µg of mouse brain tissue lysate.

Target description

~62 kDa observed.
Calculated molecular weight of 52 kDa The actual MW should be observed at ~45-65 kDa Helical membrane protein migration on SDS-PAGE may not exactly correlate with the calculated molecular weights (PNAS, 2009, 106:1760-1765).

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aritro Nath et al.
Scientific reports, 5, 14752-14752 (2015-10-02)
Hepatocellular carcinoma (HCC) is the second-leading cause of cancer-related death worldwide, and the factors influencing HCC progression are poorly understood. Here we reveal that HCC progression via induction of epithelial-mesenchymal transition (EMT) is closely associated with the expression of CD36/fatty
Cody J Aros et al.
Cell stem cell, 27(3), 413-429 (2020-07-30)
Our understanding of dynamic interactions between airway basal stem cells (ABSCs) and their signaling niches in homeostasis, injury, and aging remains elusive. Using transgenic mice and pharmacologic studies, we found that Wnt/β-catenin within ABSCs was essential for proliferation post-injury in vivo.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service