Skip to Content
Merck
All Photos(2)

Key Documents

MABS1688

Sigma-Aldrich

Anti-NCoA-2 Antibody, clone 1C6.3

clone 1C6.3, from mouse

Synonym(s):

Nuclear receptor coactivator 2, bHLHe75, Class E basic helix-loop-helix protein 75, hTIF2, NCoA-2, Transcriptional intermediary factor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
£260.00

£260.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
£260.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

£260.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C6.3, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NCOA2(10499)

General description

Nuclear receptor coactivator 2 (UniProt Q15596; also known as bHLHe75, Class E basic helix-loop-helix protein 75, hTIF2, NCoA-2, Transcriptional intermediary factor 2) is encoded by the NCOA2 (also known as BHLHE75, SRC2, TIF2) gene (Gene ID 10499) in human. Nuclear receptor coactivator 2 (NCoA-2) is a 160 kDa transcriptional coactivator for steroid receptors and nuclear receptors that acts as a coactivator of the steroid binding domain (AF-2), but not the modulating N-terminal domain (AF-1). It possesses a single NR interaction domain (NID) and two autonomous activation domains, AD1 and AD2. NCoA-2 assists nuclear receptors in the upregulation of DNA expression. In cells ligand activated nuclear receptors recruit NCoA-2 to DNA where it acylates histones to make targeted regions accessible to transcriptional machinery. Along with NCOA1 it controls energy balance between white and brown adipose tissues and also acts as a critical regulator of glucose metabolism. It is also involved in the positive regulation of the transcriptional activity of the glucocorticoid receptor NR3C1 by sumoylation enhancer RWDD3. NCoA-2 is reported to positively regulate the circadian clock by acting as a transcriptional coactivator for the CLOCK-ARNTL/BMAL1 heterodimer. Chromosomal aberrations involving NCOA2 may be a cause of acute myeloid leukemias.

Ref.: Voegel, JJ et al. (1998). EMBO J. 17, 507 519.

Specificity

Clone 1C6.3 targeted epitope lies within the C-terminal one fourth region of the full-length human NCoA-2.

Immunogen

GST-tagged recombinant human NCoA-2 C-terminal region fragment..

Application

Detect NCOA2 using this mouse monoclonal Anti-NCoA-2 Antibody, clone 1C6.3, Cat. No. MABS1688, validated for use in Immunohistochemistry (Paraffin) and Western blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected nuclear NCoA-2 immunoreactivity in human lymphoma, ovarian cancer, and small intestine tissue sections.

Quality

Evaluated by Western Blotting in human osteosarcoma cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected NCoA-2 in 10 µg of U2OS human osteosarcoma cell lysate.

Target description

~160 kDa observed. 159.0 kDa (Met1 removed) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service