MABS1145
Anti-3-PGDH Antibody, clone TK-08 13A8
clone TK-08 13A8, from mouse
Synonym(s):
D-3-phosphoglycerate dehydrogenase, 3-PGDH
Select a Size
£263.00
Estimated to ship on25 April 2025Details
Select a Size
About This Item
£263.00
Estimated to ship on25 April 2025Details
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
TK-08 13A8, monoclonal
species reactivity
human
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... PHGDH(26227)
General description
Immunogen
Application
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected 3-PGDH in human salivary gland tissue sections.
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected 3-PGDH in 10 µg of HeLa cell lysate.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service