Skip to Content
Merck
All Photos(1)

Documents

MABN450

Sigma-Aldrich

Anti-GCM2 Antibody, clone 8A10.1

clone 8A10.1, 0.5 mg/mL, from mouse

Synonym(s):

hGCMb, Chorion-specific transcription factor GCMb, GCM2, GCM 2, GCM-2, GCM motif protein 2, Glial cells missing homolog 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8A10.1, monoclonal

species reactivity

human

concentration

0.5 mg/mL

technique(s)

western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GCM2(9247)

General description

GCM2 is a nuclear protein that is involved in transcription regulation of many genes including ones important in parathyroid metabolism and development. Glial cells missing homolog 2 or GCM2 is expressed predominately in the pharyngeal pouches and, at later stages, in the developing and mature parathyroid glands. In humans, loss of GCM2 function, either through recessive apomorphic mutations or dominant inhibitor mutations in the human GCM2 gene, leads to isolated hypoparathyroidism. GCM2 expression is continued in the adult parathyroid at high levels and appears to be important in calcium sensing and calcium receptor expression in adult parathyroid cells.

Immunogen

GST-tagged recombinant protein corresponding to Human GCM2.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-GCM2 Antibody, clone 8A10.1 is validated for use in Western Blotting for the detection of GCM2.

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected GCM2 in 10 µg of SH-SY5Y cell lysate.

Target description

~ 58 kDa observed. An uncharacterized band appears at ~110 kDa in some lysates.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service