Skip to Content
Merck
All Photos(2)

Key Documents

MABN2257

Sigma-Aldrich

Anti-Nudel (NDEL1) Antibody, clone 15D5.1

clone 15D5.1, from mouse

Synonym(s):

Nuclear distribution protein nudE-like 1, Protein Nudel, Mitosin-associated protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
£332.00

£332.00


Estimated to ship on16 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
£332.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

£332.00


Estimated to ship on16 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

15D5.1, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... NDEL1(81565)

General description

Nuclear distribution protein nudE-like 1 (UniProt: Q9GZM8; also known as Protein Nudel, Mitosin-associated protein 1) is encoded by the NDEL1 (also known as EOPA, MITAP1, NUDEL) gene (Gene ID: 81565) in human. Nudel protein belongs to the nude family and is expressed in brain, heart, kidney, liver, lung, pancreas, placenta and skeletal muscle. Its expression is shown to peak during mitosis. Nudel is undergoes phosphorylation during mitosis. Three isoforms of Nudel have been reported that are produced by alternative splicing. Nudel is required for organization of the cellular microtubule array and microtubule anchoring at the centrosome. It may regulate microtubule organization at least in part by targeting the microtubule severing protein KATNA1 to the centrosome and is shown to positively regulate the activity of the minus-end directed microtubule motor protein dynein. It enhances dynein-mediated microtubule sliding by targeting dynein to the microtubule plus ends and is shown to be essential for several dynein- and microtubule-dependent processes, such as the maintenance of Golgi integrity, the centripetal motion of secretory vesicles and the coupling of the nucleus and centrosome. Nudel is required during brain development for the migration of newly formed neurons from the ventricular/subventricular zone toward the cortical plate and also plays a role in the regulation of neurite outgrowth. It localizes to the cell body of the motor neurons and colocalizes with assembled neurofilaments within axonal processes.

Specificity

Clone 15D5.1 detects Nuclear distribution protein nudE-like 1 (Nudel) in human and murine cells. It targets an epitope within 101 amino acids from the N-terminal region.

Immunogen

GST/His-taged recombinant fragment corresponding to 101 amino acids from the N-terminal region of human Nuclear distribution protein nudE-like 1 (Nudel).

Application

Anti-Nudel (NDEL1), clone 15D5.1, Cat. No. MABN2257, is a highly specific mouse monoclonal antibody that targets Nuclear distribution protein nudE-like 1 (Nudel) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Nuclear distribution protein nudE-like 1 (Nudel) in human colon, human cerebral cortex, and human tonsil tissues.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Nuclear distribution protein nudE-like 1 (Nudel) in 10 µg of mouse brain tissue lysate.

Target description

~40 kDa observed; 38.38 kda calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service