Skip to Content
Merck
All Photos(2)

Key Documents

MABN1186

Sigma-Aldrich

Anti-Prion Protein Antibody, clone 12D6.1

clone 12D6.1, from mouse

Synonym(s):

Major prion protein, ASCR, CD230, PrP, PrP27-30, PrP33-35C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12D6.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRNP(5621)

General description

Major prion protein (UniProt P04156; also known as ASCR, CD230, PrP, PrP27-30, PrP33-35C) is encoded by the PRNP (also known as ALTPRP, CJD, GSD, HDL1, KURU, PRIP, PRP, SENF) gene (Gene ID 5621) in human. The major prion protein (PrP) is synthesized with an N-terminal signal peptide (a.a. 1-22) and a C-terminal proptptide (a.a. 231-253) sequence, which are posttranslationally removed to yield the mature (a.a. 23-230) N-glycosylated, glycosylphosphatidylinositol (GPI)-anchored cell surface protein. Structurally, PrP consists of a long, disordered, flexible NH2-proximal region and a globular COOH-proximal domain linked together by a hydrophobic core (HC) region (a,a. 111-134) that is considered as a key region in both physiological and disease related processes involving the prion protein. The N-terminal region is composed of five octapeptide (PHGGGWGQ) repeats (a.a. 51-91) sandwiched between two positively charged clusters, CC1 & CC2 (a.a. 23-27 & 95-110). The exact physiological function of PrP is not known, although various biological functions have been suggested for this protein, including signal transduction, neurotransmitter metabolism, cell adhesion, antioxidant activity, neurogenesis, immune cell activation, copper metabolism and homeostasis of trace elements. Transmissible spongiform encephalopathies (TSEs) represent a family of rare and fatal neurodegenerative disorders, including Creutzfeldt-Jakob disease, Gerstmann-Sträussler-Scheinker syndrome, fatal familial insomnia and kuru in human, bovine spongiform encephalopathy in cattle, and scrapie in sheep. It is widely accepted that the conformational transition of the native and predominantly α-helical cellular PrP (PrPC) to a β-sheet-rich pathogenic scrapie isoform (PrPSc) is responsible for the accumulation of PrPSc aggregates.

Specificity

Clone 12D6.1 detects an epitope present in human PrP and the spliced isoform PrP(M8) reported by UniProt (P04156-1 & P04156-2), but absent in the alternative prion protein AltPrP (UniProt F7VJQ1-1).

Immunogen

Epitope: Internal (N-terminal half).
Human IgG1 Fc-tagged recombinant human prion protein internal fragment.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected prion protein in human cerebral cortex tissue sections.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-Prion Protein Antibody, clone 12D6.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of Prion Protein.

Quality

Evaluated by Western Blotting of recombinant human prion protein.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected 0.05 µg of human IgG1 Fc-tagged recombinant human prion protein internal fragment.

Target description

27.66/26.89 kDa (PrP/PrP(M8) prepro-form), 22.75/25.24 kDa (mature/pro-form) calculated.

Linkage

Replaces: MAB1562

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service