Skip to Content
Merck
All Photos(1)

Key Documents

MABF1107

Sigma-Aldrich

Anti-DDX41 Antibody, clone 4D8.2

clone 4D8.2, from mouse

Synonym(s):

DEAD box protein 41, DEAD box protein abstrakt homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D8.2, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... DDX41(51428)

General description

Probable ATP-dependent RNA helicase DDX41 (UniProt: Q9UJV9; also known as DEAD box protein 41; DEAD box protein abstrakt homolog) is encoded by DDX41 (also known as ABS) gene (Gene ID 51428) in human. DDX41 belongs to the family of DEAD box helicase family of proteins, characterized by the presence of the conserved Asp-Glu-Ala-Asp (DEAD) motif. DDX41 is reported to sense DNA virus in myeloid dendritic cells (mDCs) mediated by the adaptor STING (stimulator of interferon genes). DDX41 is reported to bind to both DNA and STING through a DEADc domain and then DDX41-STING complex localizes to the cytoplasm. DDX41 binds bacterial DNA in the infected cells and is shown to be more important than gamma-interferon-inducible protein IFI16 in the initial sensing of cytosolic B-form DNA in triggering the early burst of type I interferon response. Knockdown of DDX41 expression blocks the ability of mDCs to mount type I interferon and cytokine responses to DNA and DNA viruses. Ref.: Zhang, Z., et al. (2011). Nat. Immunol. 12(10): 959-966.

Specificity

Clone 4D8.2 detects DDX41 protein in HEPG2 cells by Western blotting and in human ovarian and cervical cancer tissue by immunohisotchemistry.

Immunogen

GST-tagged recombinant fragment corresponding to 115 amino acids from the N-terminal region of human DDX41.

Application

Research Category
Inflammation & Immunology
This mouse monoclonal Anti-DDX41 antibody, clone 4D8.2, Cat. No. MABF1107 is validated for use in Immunohistochemistry (Paraffin) and Western Blotting for the detection of DDX41.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected DDX41 in 10 µg of HeLa cell lysate.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected DDX41 in human ovarian cancer, human cervical cancer, and human prostate cancer tissue.




Quality

Evaluated by Western Blotting in HEPG2 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected DDX41 in 10 µg of HEPG2 cell lysate

Target description

~70 kDa observed; 69.8 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service