Skip to Content
Merck
All Photos(1)

Key Documents

MABE1990

Sigma-Aldrich

Anti-PLZF/ZBTB16 Antibody, clone Mags-21F7

Synonym(s):

Promyelocytic leukemia zinc finger protein, Zinc finger and BTB domain-containing protein 16, Zinc finger protein 145, Zinc finger protein PLZF

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
£144.00
100 μG
£362.00

£144.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
£144.00
100 μG
£362.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

£144.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

biological source

hamster

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Mags-21F7, monoclonal

mol wt

observed mol wt ~75 kDa

purified by

using protein G

species reactivity

human, mouse

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... ZBTB16(7704)

Specificity

Clone Mags-21F7 is an Armenian Hamster monoclonal antibody that detects Zinc finger and BTB domain-containing protein 16 (PLZF/ZBTB16).

Immunogen

A combination of peptides covering the amino, carboxyl, and hinge regions of human Zinc finger and BTB domain-containing protein 16 (PLZF/ZBTB16).

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant Human PLZF/ZBTB16 protein.

Western Blotting Analysis: A 1:500 dilution of this antibody detected full-length, FLAG-tagged recombinant Human PLZF/ZBTB16.

Tested Applications

Western Blotting Analysis: A representative lot detected PLZF/ZBTB16 in Western Blotting applications (Kovalovsky, D., et al. (2008). Nat Immunol. 9(9):1055-64).

Flow Cytometry Analysis: A representative lot detected PLZF/ZBTB16 in Flow Cytometry applications (Kovalovsky, D., et al. (2008). Nat Immunol.;9(9):1055-64; Kovalovsky, D., et al. (2010). J Immunol. 184(12):6746-55).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Zinc finger and BTB domain-containing protein 16 (UniProt: Q05516; also known as Promyelocytic leukemia zinc finger protein, Zinc finger protein 145, Zinc finger protein PLZF) is encoded by the ZBTB16 (also known as PLZF, ZNF145) gene (Gene ID: 7704) in human. PLZF is a member of the krueppel C2H2-type zinc-finger protein family with nine C2H2-tpe zinc-finger domains. It serves as a transcriptional repressor and this repression may be mediated through recruitment of histone deacetylases to target promoters. It affects diverse signaling pathways, including growth-regulatory and differentiation pathways and is a key regulator of myeloid development. Its expression is observed in early, but in differentiated hematopoietic cells. It is expressed in bone marrow, early myeloid cells, and peripheral blood mononuclear cells. At lower level it is also detected in the ovary, kidney, and lung tissue. PLZF is reported to be essential for development of the characteristic innate/effector functions of natural killer T (NKT) cells. It is highly expressed during the differentiation of NKT cells and is essential for the acquisition of effector/memory innate-like functions. PLZF-deficient NKT cells lose their characteristic memory phenotype, and their function is significantly impaired. These cells lose their unique ability to co-secrete IL-4 and IFN- upon ex vivo stimulation. (Ref.: Kolesnichenko, M., and Vogt, PK. (2011). Cell Cycle.10(5); 771-775; Kovalovsky, D., et al. (2010). J. Immunol. 184: 6746-6755; Melnick, AM., et al. (2000). Mol. Cell Biol. 20(6); 2075-2086).

Physical form

Purified Armenian hamster monoclonal antibody IgG2-IgG3 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service