Skip to Content
Merck
All Photos(1)

Documents

MABD20

Sigma-Aldrich

Anti-PAX7 Antibody, clone 2F12H4

clone 2F12H4, from rat

Synonym(s):

paired box 7, PAX7 transcriptional factor, paired box gene 7, paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F12H4, monoclonal

species reactivity

mouse, human (Harada, A., et al. (2009). Hybridoma, 26:6. Hyb.2009.0039)

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Pax7(18509)

General description

Paired box protein Pax-7 (PAX7) belongs to the Pax family of transcription factors, a highly conserved group of proteins with crucial roles in tissue homeostasis and organogenesis. PAX7 is involved in the specification, survival, maintenance, and self-renewal of skeletal muscle progenitors and myogenic progenitors. It is primarily expressed in newly activated and quiescent satellite cells. Studies have shown that on activation, quiescent satellite cells co-express MyoD with PAX7 prior to proliferation, after which one or the other transcription factor is downregulated. Downregulation of MyoD instead of PAX7 leads to withdrawal from the cell cycle and myogenic differentiation, causing the cell to return to seemingly quiescent state. Chromosomal aberrations relating to PAX7 have been linked to rhabdomysarcoma 2 (RMS2).

Immunogen

KLH-conjugated linear peptide corresponding to mouse PAX7.

Application

Anti-PAX7 Antibody, clone 2F12H4 is a Rat monoclonal antibody for detection of PAX7 also known as paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7 & has been validated in WB, ICC.
Immunocytochemistry: A previous lot of this antibody was shown to detect Pax7 localized in euchromatin of human Ewing’s
sarcoma cells. The antibodies are detected with Alexa Fluor 488-conjugated anti-rat IgG. Each sample is counterstained with hoechst (Harada, A., et al. (2009). Hybridoma, 26:6. Hyb.2009.0039).
Research Category
Stem Cell Research
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in mouse fetal brain tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected PAX7 in 10 µg of mouse fetal brain tissue lysate.

Target description

~ 55 kDa observed.
The calculated MW of PAX7 is 55 kDa; however, expression of PAX7 can have multiple isoforms. (Robert B., et al. (2008). Physiol. Genomics 33:41-49.)

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse fetal brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luthfia Dewi et al.
Food & function, 15(8), 4010-4020 (2024-03-19)
Cordyceps sinensis is a parasitic fungus known to induce immune responses. The impact of Cordyceps supplementation on stem cell homing and expansion to human skeletal muscle after exercise remains unexplored. In this study, we examined how pre-exercise Cordyceps supplementation influences

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service