MAB844-2
Anti-Parainfluenza 2 Antibody, clone 233/9A
ascites fluid, clone 233/9A, Chemicon®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
233/9A, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunofluorescence: suitable
isotype
IgG1
suitability
not suitable for immunoprecipitation
shipped in
dry ice
Specificity
Specific for the HN protein common to all parainfluenza II strains. No cross-reactivity to Parainfluenza I, III or IV.
Immunogen
Greer
Application
Detect Parainfluenza 2 using this Anti-Parainfluenza 2 Antibody, clone 233/9A validated for use in ELISA & IF.
ELISA: 1:800+.
Indirect Immunofluorescence: 1:100-200+ (fresh frozen tissue sections).
Optimal working dilutions must be determined by end user.
Not suggested for use in immunoprecipitation.
Indirect Immunofluorescence: 1:100-200+ (fresh frozen tissue sections).
Optimal working dilutions must be determined by end user.
Not suggested for use in immunoprecipitation.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Storage and Stability
Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service