Skip to Content
Merck
All Photos(1)

Documents

MAB1944-C

Sigma-Aldrich

Anti-Collagen VI alpha-3 Antibody, clone 3C4, Ascites Free

culture supernatant, clone 3C4, from mouse

Synonym(s):

Collagen alpha-3(VI) chain, Collagen VI alpha-3 polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

3C4, monoclonal

species reactivity

human

technique(s)

electron microscopy: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... COL6A3(1293)

Related Categories

General description

Collagen alpha-3(VI) chain (UniProt P12111; also known as Collagen VI alpha-3 polypeptide) is encoded by the COL6A3 gene (Gene ID 1293) in human. Type VI collagen is an extracellular matrix (ECM) component present in virtually all connective tissues, including cartilage, bone, tendon, muscles and cornea, where it forms microfibrils in close association with basement membranes. In addition to anchoring the basement membrane to the pericellular matrix in muscle, research also indicates a role for collagen VI in cell signaling and cell migration. The basic structural unit of collagen VI is a heterotrimer composed of the alpha-1(VI), alpha-2(VI), and alpha-3(VI) chains (encoded by the COL6A1, COL6A2, and COL6A3 genes, respectively). The α1(VI) and α2(VI) chains are similar in size and domain structure, they contain a 335- or 336-amino acid triple helix region that is characteristic of all collagens. Flanking the triple helix are domains homologous to the A-type domains found in von Willebrand factor (VWA domains). α1(VI) and α2(VI) contain one VWA domain N-terminal to the triple helix (N1) and two VWA domains C-terminal of the helix (C1 and C2). The α3(VI) chain, on the other hand, is much larger with 10 N-terminal (N1–N10) and two C-terminal VWA domains (C1 and C2), and several other types of identifiable domains in the C terminal region (C3–C5). Mutations in the COL6A1, COL6A2, and COL6A3 genes are known causes of Ullrich congenital muscular dystrophy (UCMD) and Bethlem myopathy (BM). Three additional type VI collagen chains have been reported in 2008 (α4(VI), α5(VI) and α6(VI) chains encoded by COL6A4, COL6A5, and COL6A6, respectively).

Immunogen

Purified human collagen VI.

Application

This Anti-Collagen VI alpha-3 Antibody, clone 3C4, Ascites Free is validated for use in Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Electron Microscopy for the detection of Collagen VI alpha-3.

Quality

Evaluated by Immunohistochemistry in human breast tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected collagen VI alpha-3 chain immunoreactivity in human breast tissue section.

Target description

343.7/321.4/113.2/278.2/134.7 kDa (isoform 1/2/3/4/5 pro-form) and 340.8/318.5/110.4/275.3/131.8 kDa (isoform 1/2/3/4/5 mature form) calculated

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service