Skip to Content
Merck
All Photos(1)

Documents

MAB1304

Sigma-Aldrich

Mouse Anti-Human IgG Antibody, Fd, clone HP6045

clone HP6045, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

HP6045, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Specificity

Human IgG Fd.

ELISA Specificity: (Hamilton et al., 1987)

human IgG subclass 1 100%

human IgG subclass 2 100%

human IgG subclass 3 100%

human IgG subclass 4 99%

human IgG Pan 100%

human IgG Fc 0.04%

human IgG Fab 100%



Isoelectric Focusing: mean pI 6.5

Immunogen

Epitope: Fd region

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies
This Mouse anti-Human IgG Antibody, Fd, clone HP6045 is validated for use in ELISA, RIA, WB for the detection of Human IgG.
Useful as both a capture antibody and a detection antibody for human IgG Fd (Papadea, C., et al., Clin. Chem. (1985). 31(12):1940-1945; Hamilton, R.G., et al., Electrophoresis (1987) 8:127-134).

ELISA

Radioimmunoassay (RIA)

Western blotting.

Final working dilutions must be determined by end user.

Physical form

Format: Purified
Liquid in 0.02 M phosphate buffer with 0.25 M NaCl and 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to six months after date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shohei Ogata et al.
PloS one, 8(12), e81448-e81448 (2013-12-11)
Although intravenous immunoglobulin (IVIG) is highly effective in Kawasaki disease (KD), mechanisms are not understood and 10-20% of patients are treatment-resistant, manifesting a higher rate of coronary artery aneurysms. Murine models suggest that α2-6-linked sialic acid (α2-6Sia) content of IVIG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service