Skip to Content
Merck
All Photos(2)

Key Documents

HTS091M

Sigma-Aldrich

ChemiSCREEN Human DP Prostanoid Receptor Membrane Preparation

Human DP GPCR membrane preparation for Radioligand binding Assays & GTPgammaS binding.

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
£668.00

£668.00


Please contact Customer Service for Availability


Select a Size

Change View
200 UNITS
£668.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.41

£668.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... PTGDR(5729)

General description

Full-length human PTGDR cDNA encoding DP
Prostanoids are a series of arachidonic acid metabolites produced by the action of cyclooxygenase and subsequently by isomerases and synthases. Cells rapidly secrete prostanoids after synthesis, whereupon the prostanoids bind to a family of 8 GPCRs to exert their biological effects (Narumiya and FitzGerald, 2001). The prostaglandin PGD2 is produced by mast cells upon activation by allergens, and is present at high levels in allergic diseases. PGD2 binds to two receptors, DP and CRTH2. DP activates Gs to increase cAMP levels, and lack of DP results in reduced allergic response in animal models of bronchial asthma (Matsuoka et al., 2000). Chemicon′s DP membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of DP interactions and its ligands. The membrane preparations exhibit a Kd of 10.39 nM for [3H]-PGD2. With 8 nM [3H]-PGD2, 5 µg/well and 10 µg/well DP Membrane Prep typically yield greater than 4-fold signal-to-background ratio.

Application

Radioligand binding assay and GTPS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: DP
Target Sub-Family: Prostanoid

Quality

Signal:background and specific binding values obtained in a competition binding assay with varying amounts of DP membrane prep:
10 µg/well5 µg/well
Signal:Background 4.86 4.12
Specific Binding (cpm) 1345 885



Table 2. IC50 values for ligands obtained in a competition binding assay with DP membrane preparation.
IC50 (nM)
Prostaglanding D2 12.1
13,14-dihydro-15-keto prostaglandin D210000
BW A868C4.8


SPECIFICATIONS: 1 unit = 10 µg
Bmax for [3H] PGD2 binding: 5.3 pmol/mg protein;
Kd for [3H] PGD2 binding: ~10.39nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [3H] PGD2 (Perkin Elmer#:NET-616 )
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 4-fold signal:background with 3H-labeled PGD2 at 8 nM

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.
Packaging method: Membranes protein were adjusted to 0.5 mg/ml in 1 ml packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Matsuoka et al.
Science (New York, N.Y.), 287(5460), 2013-2017 (2000-03-17)
Allergic asthma is caused by the aberrant expansion in the lung of T helper cells that produce type 2 (TH2) cytokines and is characterized by infiltration of eosinophils and bronchial hyperreactivity. This disease is often triggered by mast cells activated
Genetic and pharmacological analysis of prostanoid receptor function.
S Narumiya et al.
The Journal of clinical investigation, 108(1), 25-30 (2001-07-04)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service