HTS082RTA
Ready-to-Assay 5-HT 2A Serotonin Family Receptor Frozen Cells
Human
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.
5-Hydroxytryptamine (5-HT, also commonly known as serotonin) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals. In the periphery, 5-HT mediates gastrointestinal motility, platelet aggregation, and contraction of blood vessels. Many functions of the central nervous system are influenced by 5-HT, including sleep, motor activity, sensory perception, arousal and appetite. A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT (Hoyer et al., 1994). 5-HT2A, which couples to Gq/11 to increase intracellular calcium, is widely expressed at central and peripheral sites of 5-HT action, and contributes to many of the physiological effects of 5-HT. The hallucinogenic activity of LSD is mediated in part by its action as an partial to full agonist at 5-HT2A, and the activity of atypical antipsychotics such as clozapine appears to be mediated in part by antagonism of 5-HT2A (Barnes and Sharp, 1999). Millipore′s cloned human 5-HT2A -expressing cell line is made in the Chem-1 host, which supports high levels of recombinant 5-HT2A expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at 5-HT2A.
5-Hydroxytryptamine (5-HT, also commonly known as serotonin) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals. In the periphery, 5-HT mediates gastrointestinal motility, platelet aggregation, and contraction of blood vessels. Many functions of the central nervous system are influenced by 5-HT, including sleep, motor activity, sensory perception, arousal and appetite. A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT (Hoyer et al., 1994). 5-HT2A, which couples to Gq/11 to increase intracellular calcium, is widely expressed at central and peripheral sites of 5-HT action, and contributes to many of the physiological effects of 5-HT. The hallucinogenic activity of LSD is mediated in part by its action as an partial to full agonist at 5-HT2A, and the activity of atypical antipsychotics such as clozapine appears to be mediated in part by antagonism of 5-HT2A (Barnes and Sharp, 1999). Millipore′s cloned human 5-HT2A -expressing cell line is made in the Chem-1 host, which supports high levels of recombinant 5-HT2A expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at 5-HT2A.
Cell Line Description
- GPCR Cell Lines
- Host cells: Chem-1
Application
Human 5-HT2A GPCR frozen cells for Calcium Flux FLIPR Assays.
Biochem/physiol Actions
- GPCR Class: A
- Protein Target: 5-HT2A
- Target Sub-Family: Serotonin
Components
Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.
Fifty (50) mL of Media Component.
Disclaimer
This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service