Skip to Content
Merck
All Photos(1)

Key Documents

HTS016RTA

Sigma-Aldrich

Ready-to-Assay C3a Anaphylotoxin Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
£696.00

£696.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
£696.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

£696.00


Please contact Customer Service for Availability

Product Name

Ready-to-Assay C3a Anaphylotoxin Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

C3a, along with C4a and C5a, is a 77 amino acid anaphylotoxin generated by proteolytic cleavage during activation of the complement pathway (Ember and Hugli, 1997). The anaphylotoxins strongly promote inflammation by recruiting leukocytes, particularly basophils, eosinophils, neutrophils and monocyte (Martin et al., 1997). The proinflammatory properties of C3a are mediated by interaction between the peptide and a 7-TM G-protein coupled receptor, C3aR (Crass et al., 1996). Genetic and pharmacological inhibition of C3a/C3aR interaction indicates an important role for C3aR in allergic asthma (Humbles et al., 2000; Drouin et al., 2002). C3a/C3aR also enhances the effect of SDF-1 in promoting retention of haematopoietic stem/progenitor cells within the bone marrow (Rataiczak et al., 2004). Millipore′s cloned human C3a-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant C3a expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists, and modulators at C3a.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human C3aR GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: C3aR
  • Target Sub-Family: Anaphylotoxin

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service