Skip to Content
Merck
All Photos(1)

Key Documents

GF180

Sigma-Aldrich

MIF,
Human Recombinant

Synonym(s):

Macrophage Migration Inhibitory Factor

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
£300.00

£300.00


Estimated to ship on16 April 2025Details



Select a Size

Change View
25 μG
£300.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

£300.00


Estimated to ship on16 April 2025Details


impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

dry ice

General description

Macrophage Migration Inhibitory Factor (MIF) is a small secreted protein that can act as a pleiotropic pro-inflammatory cytokine as well as an enzyme. MIF pro-inflammatory activity can be initiated by signaling through CD74 and CD44, resulting in the secretion of TNF-a, IL-1, IL-6, IL-8, and various MMPs. The enzymatic activity of MIF is characterized by its ability to act as a tautomerase, capable of catalyzing the keto to enol isomerization of keto-phenylpyruvate and L-dopachrome. It appears as though MIF catalytic activity is dependent upon a trimeric configuration and a free N-terminal proline residue.
Insect cell derived recombinant MIF is a 15 kDa protein containing 124 amino acid residues, including an N-terminal His-tag.
Product Source: Protein is expressed in insect cells.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

Determined by its ability to inhibit monocyte migration.

Physical form

Product is presented in 10mM Sodium Citrate, pH 3.0 and is filtered through a 0.2 micron filter before lyophilization.

Storage and Stability

Store at -20°C for up to 4 months from date of receipt Centrifuge the vial prior to opening.
Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein and store in working aliquots at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service