Skip to Content
Merck
All Photos(1)

Documents

CP1ALLRES

Milli-Q

AFS® Cartridge

For AFS® 3 systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
NA.21

Quality Level

feature

Virgin ion-exchange resins for ion removal

packaging

pkg of 1 unit

manufacturer/tradename

AFS®

resistivity

>18.0 MΩ-cm (at 25 °C)

compatibility

for use with AFS® (1999-2012)

shipped in

ambient

General description

AFS® Cartridges contain a combination of purification media carefully selected to remove trace ionic contaminants affecting specific applications. Cartridges are constructed from polypropylene and polyethylene, which are suitable for the production of pure water and are qualified for low organics and ionic extractables.

Application

The AFS® Cartridge removes ionic contaminants to match the water quality required for your applications.

Features and Benefits

  • Guarantee water quality.
  • Ensure compliance with system specifications.
  • Purification technology involves the use of ion-exchange resins for ion removal.

Components

Virgin ion-exchange resins for ion removal

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service